EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Design, synthesis and DNA-cleavage of Gly-Gly-His-naphthalene diimide conjugates.
PMID 10571151
Abstract
Naphthalene diimides with one and two metal-chelating Gly-Gly-His (GGH) motifs have been synthesized. Both conjugates induce single-stranded cleavage of plasmid pBR322 DNA in the presence of nickel and Oxone and are approximately 100-fold more efficient than Ni(II) x GH-CONH2 itself.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G4541 Gly-Gly-His
C10H15N5O4