EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Regiospecific internal desaturation of aliphatic compounds by a mutant Rhodococcus strain.
PMID 10584034
Abstract
A mutant Rhodococcus strain lacking the ability to utilize 1-chlorohexadecane was found to cis-desaturate aliphatic compounds, such as 1-chlorohexadecane, n-hexadecane, and heptadecanonitrile, yielding corresponding products with a double bond mainly at the ninth carbon from the terminal methyl groups. A new oxidative pathway involving the cis-desaturation step was suggested for alkane utilization by Rhodococcus spp.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
245623
1-Chlorohexadecane, 95%
C16H33Cl
24750
1-Chlorohexadecane, purum, ≥97.0% (GC)
C16H33Cl