EMAIL THIS PAGE TO A FRIEND
Natural product reports
The discovery of a mevalonate-independent pathway for isoprenoid biosynthesis in bacteria, algae and higher plants.
PMID 10584331
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
95098
(E)-1-Hydroxy-2-methyl-2-butenyl 4-pyrophosphate lithium salt, ≥95.0% (TLC)
C5H12O8P2 · xLi+