EMAIL THIS PAGE TO A FRIEND
Biotechnology progress
Membrane chromatography: preparation and applications to protein separation.
PMID 10585183
Abstract
As a result of the convective flow of solutes through porous membranes, membrane chromatography has a higher capture efficiency and a higher productivity than column chromatography and shows most promising industrial applications for the recovery, isolation, and purification of proteins and enzymes. This paper presents a comprehensive review of the methods for preparation of adsorptive membranes (such as surface modification, in situ copolymerization, direct formation from hydrophilic materials, and functionalized particulate-entrapped membranes) and deals particularly with novel macroporous chitin and chitosan membranes for protein separations developed by the authors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S4027 Sulfoxyethyl cellulose Fast Flow, fibers