EMAIL THIS PAGE TO A FRIEND
Teratogenesis, carcinogenesis, and mutagenesis
Inhibitory effect of propolis and bee venom on the mutagenicity of some direct- and indirect-acting mutagens.
PMID 10587410
Abstract
The antimutagenic effect of ethanolic extract of propolis (EEP) and honeybee (Apis mellifera) venom, both collected in the State of São Paulo, Brazil, was assessed by the Salmonella/microsome assay upon direct- and indirect-acting mutagens. EEP had inhibitory effect (in an ascending order) on the mutagenicity power of daunomycin (TA102), benzo(a)pyrene (TA100), and aflatoxin B(1)(TA98) and the venom acted against the mutagenicity of 4-nitro-o-phenylenediamine (TA98) and daunomycin (TA102). Teratogenesis Carcinog. Mutagen. 19:403-413, 1999.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
108898
4-Nitro-o-phenylenediamine, 98%
C6H7N3O2