EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Degradation of the basic helix-loop-helix/Per-ARNT-Sim homology domain dioxin receptor via the ubiquitin/proteasome pathway.
PMID 10593927
Abstract
The basic helix-loop-helix/Per-ARNT-Sim homology domain dioxin receptor (DR) translocates to the nucleus upon binding of aromatic hydrocarbon ligands typified by dioxin, whereupon it partners the Ah receptor nuclear translocator and initiates transcription. Concurrently, ligand binding down-regulates receptor levels via an unknown mechanism. In this study we show that receptor levels are dependent upon cellular compartmentalization, with entry into the nucleus leading to the rapid destruction of the DR. Ligand-induced DR translocation was bypassed by adding a heterologous nuclear localization signal to the DR, creating a constitutively nuclear form of the dioxin receptor (DRNLS). The DRNLS protein was shown to be unstable with a half-life of