EMAIL THIS PAGE TO A FRIEND
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists
Acetohexamide-acetazolamide mix-up during emergency treatment.
PMID 10595808
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A178
Acetohexamide, analytical standard
C15H20N2O4S