EMAIL THIS PAGE TO A FRIEND
Seminars in oncology
Clinical pharmacokinetics of antitumor antifolates.
PMID 10598559
Abstract
Antifolate drugs, as a class, have broad-spectrum activity against both hematologic and solid human malignancies. The pharmacokinetics of the classical antifolate methotrexate have been well-defined and pharmacokinetic data can be exploited to reduce the toxicity and enhance the activity of the drug. Methotrexate remains the only anticancer drug for which plasma drug level monitoring is used in routine clinical practice. Recently, novel classical and nonclassical antifolates have been developed that target either specific folate-dependent enzymes (e.g., thymidylate synthase [CB3717, raltitrexed, ZD9331, 1843U89, nolatrexed, AG331], glycinamide ribonucleotide transformylase [lometrexol, LY309887, AG2034] or multiple folate-dependent enzymes (e.g., MTA/LY231514). In the early clinical trials of these agents, a number of pharmacokinetic-pharmacodynamic relationships were identified and it is highly likely that the full therapeutic potential of these new drugs will also require the exploitation of pharmacokinetic data.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0040
Lometrexol hydrate, ≥95% (HPLC)
C21H25N5O6 · xH2O