EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Hypernuclear acetylation in atherosclerotic lesions and activated vascular smooth muscle cells.
PMID 10600518
Abstract
Recent studies have implicated acetylation of several nuclear proteins such as histones and p53 on their epsilon-portion of lysine residues in eukaryotic transcription. Here we raised a specific polyclonal antibody against epsilon-acetylated lysine. Using the antibody, we detected hypernuclear acetylation (HNA) in atherosclerotic vascular smooth muscle cells (VSMCs). Thrombin, a humoral factor known to cause activation and proliferation of VSMCs, strongly potentiated HNA in cultured VSMCs. MAP kinase pathway and a signal coactivator CREB binding protein (CBP) were involved in thrombin-induced HNA of VSMCs. Our results suggest that coactivators cooperating with signal-dependent transcription activators play an important role in atherosclerogenesis via HNA in VSMCs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4021
Nε-Acetyl-L-lysine
C8H16N2O3