EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Activities of synthetic hybrid peptides against anaerobic bacteria: aspects of methodology and stability.
PMID 10602725
Abstract
The increasing problem of antibiotic resistance among pathogenic bacteria requires development of new antimicrobial agents. One line of investigation is the synthesis of antimicrobial hybrid peptides. The aim of the present investigation was to determine the in vitro activities of 16 cecropin-melittin hybrid peptides (CAMEL analogues) against 60 anaerobic bacterial strains, to compare their activities with those of seven clinically used antimicrobial agents, and to compare different methods for anaerobic susceptibility testing of these peptides. The stability of one of the peptides, temporin B, with different stereoisomeric configurations was investigated in a fecal milieu. The CAMEL analogues showed antimicrobial activity against the anaerobic bacteria, with MICs ranging from 0.125 to 32 microg/ml. The overall activities (the MICs at which 90% of isolates are inhibited) of the CAMEL analogues against anaerobic bacteria were mainly inferior to those of imipenem, clindamycin, and piperacillin but were equal to or superior to those of metronidazole, cefoxitin, ciprofloxacin, and chloramphenicol. The agarose dilution method was found to be an accurate method for the testing of large numbers of bacterial strains. The D isomer of temporin B was inactivated more slowly in feces than the L isomer. This study shows that the CAMEL analogues are potential agents for the treatment of anaerobic infections.