EMAIL THIS PAGE TO A FRIEND
The Journal of infectious diseases
Risk of influenza A (H5N1) infection among health care workers exposed to patients with influenza A (H5N1), Hong Kong.
PMID 10608786
Abstract
The first outbreak of avian influenza A (H5N1) occurred among humans in Hong Kong in 1997. To estimate the risk of person-to-person transmission, a retrospective cohort study was conducted to compare the prevalence of H5N1 antibody among health care workers (HCWs) exposed to H5N1 case-patients with the prevalence among nonexposed HCWs. Information on H5N1 case-patient and poultry exposures and blood samples for H5N1-specific antibody testing were collected. Eight (3.7%) of 217 exposed and 2 (0.7%) of 309 nonexposed HCWs were H5N1 seropositive (P=.01). The difference remained significant after controlling for poultry exposure (P=.01). This study presents the first epidemiologic evidence that H5N1 viruses were transmitted from patients to HCWs. Human-to-human transmission of avian influenza may increase the chances for the emergence of a novel influenza virus with pandemic potential.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500928 Anti-AVIAN INFLUENZA A M2 antibody produced in rabbit, affinity isolated antibody
SAB3500924
Anti-AVIAN INFLUENZA HEMAGGLUTININ 2 antibody produced in rabbit, affinity isolated antibody
SAB3500925
Anti-AVIAN INFLUENZA HEMAGGLUTININ 3 antibody produced in rabbit, affinity isolated antibody
SAB3500907 Anti-AVIAN INFLUENZA HEMAGGLUTININ antibody produced in rabbit, affinity isolated antibody
SAB3500908
Anti-AVIAN INFLUENZA HEMAGGLUTININ antibody produced in rabbit, affinity isolated antibody
SAB3500905 Anti-AVIAN INFLUENZA NEURAMINIDASE antibody produced in rabbit, affinity isolated antibody
SAB3500906 Anti-AVIAN INFLUENZA NEURAMINIDASE antibody produced in rabbit, affinity isolated antibody
SAB3500869 Monoclonal Anti-HEMAGGLUTININ antibody produced in mouse, clone 4F1F2, affinity isolated antibody
SAB3500870 Monoclonal Anti-HEMAGGLUTININ antibody produced in mouse, clone 4H1C10, affinity isolated antibody
SAB3500871
Monoclonal Anti-HEMAGGLUTININ antibody produced in mouse, clone 1E6A7, affinity isolated antibody
SAB3500872
Monoclonal Anti-HEMAGGLUTININ antibody produced in mouse, clone 1E7D8, affinity isolated antibody
SAB3500873
Monoclonal Anti-HEMAGGLUTININ antibody produced in mouse, clone 7B9B2, affinity isolated antibody
SAB3500874
Monoclonal Anti-HEMAGGLUTININ antibody produced in mouse, clone 4E10C10, affinity isolated antibody
SAB3500875
Monoclonal Anti-HEMAGGLUTININ antibody produced in mouse, clone 4E11E1, affinity isolated antibody