EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
Cytotoxic constituents from Solidago virga-aurea var. gigantea MIQ.
PMID 10615872
Abstract
Activity-guided fractionation of the whole plant of Solidago virga-aurea var. gigantea M(IQ). (Compositae) has led to the isolation of three cytotoxic compounds, erythrodiol-3-acetate (1), alpha-tocopherol-quinone (2), and trans-phytol (3) from the hexane soluble fraction. It is the first report of those compounds from the genus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09258
Erythrodiol, analytical standard
C30H50O2