EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Stereochemical differentiation of cyclic glycols by ion-molecule reactions in a quadrupole mass spectrometer using dimethyl ether acetonitrile and 2-S-pyrrolidinemethanol
PMID 10623933
Abstract
In this study, ion-molecule reactions using chemical ionization in the positive ion mode using dimethyl ether, acetonitrile and 2-S-pyrrolidinemethanol as reagent gases have been used to distinguish between cis- and trans-1,2-cyclopentanediol and cis- and trans-1,2-cyclohexanediol. In the gas phase, the stereospecific ion-molecule reaction gives characteristic ions and substantial differences are observed in their relative abundances from which the diastereoisomers of those cyclic glycols can be clearly differentiated. Copyright 2000 John Wiley & Sons, Ltd.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
361445
(±)-trans-1,2-Cyclopentanediol, 97%
C5H10O2