EMAIL THIS PAGE TO A FRIEND
Nuclear medicine and biology
Radioiodinated antibody targeting of the HER-2/neu oncoprotein: effects of labeling method on cellular processing and tissue distribution.
PMID 10628557
Abstract
Monoclonal antibody (MAb) internalization can have a major effect on tumor retention of radiolabel. Two anti-HER-2/neu MAbs (TA1 and 520C9) were radioiodinated using the iodogen, N-succinimidyl 5-iodo-3-pyridinecarboxylate (SIPC), and tyramine-cellobiose (TCB) methods. Paired-label studies compared internalization and cellular processing of the labeled MAbs by SKOv3 9002-18 ovarian cancer cells in vitro. Intracellular radioiodine activity for 520C9 was up to 2.6 and 3.0 times higher for SIPC and TCB labeling, respectively, compared with iodogen. Likewise, intracellular activity for TA1 was up to 2.3 and 2.9 times higher with the SIPC and TCB methods compared with iodogen labeling. Unfortunately, similar advantages in tumor accumulation were not achieved in athymic mice bearing SKOv3 9008-18 ovarian cancer xenografts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0656
1,3,4,6-Tetrachloro-3α,6α-diphenylglycouril
C16H10Cl4N4O2