EMAIL THIS PAGE TO A FRIEND
Archives of environmental contamination and toxicology
Effect of different combinations of MGK-264 or piperonyl butoxide with plant-derived molluscicides on snail reproduction.
PMID 10629280
Abstract
Effect of sublethal treatment (20% and 60% of LC(50)/24 h) of plant-derived molluscicides, viz. Polianthes tuberosa, Trachyspermum ammi, Allium sativum powder; Azadirachta indica oil; oleoresin of Zingiber officinale; and their active molluscicidal component in combination (1:5) with MGK-264 or piperonyl butoxide on the reproduction of snail Lymnaea acuminata have been studied. It was observed that the combination of plant derived molluscicide and their active molluscicidal components, viz. tigogenin, hecogenin, azadirachtin, allicin, thymol, and [6]-gingerol combination with MGK-264 or piperonyl butoxide caused a significant reduction in fecundity, hatchability, and survival of young snails. Withdrawal of snails to fresh water after the above treatment caused a significant recovery in the fecundity of L. acuminata.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36168
MGK 264, PESTANAL®, analytical standard
C17H25NO2