EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Discovery of orally active nonpeptide vitronectin receptor antagonists based on a 2-benzazepine Gly-Asp mimetic.
PMID 10633035
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
385409
4-Bromo-3-methylanisole, 98%
C8H9BrO