EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
Chiral 2-amino-1-butanol xanthone derivatives as potential antiarrhythmic and hypotensive agents.
PMID 10635353
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A43804
2-Amino-1-butanol, 97%
C4H11NO