EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of urology and nephrology
Tiaprofenic acid-induced cystitis--three cases and a literature review.
PMID 10636583
Abstract
Tiaprofenic acid is a non-steroidal anti-inflammatory drug that may cause severe non-bacterial cystitis. Three cases are described and the literature is reviewed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1410900 Tiaprofenic acid, European Pharmacopoeia (EP) Reference Standard
C14H12O3S