The Journal of biological chemistry
Glucosidase and mannosidase inhibitors mediate increased secretion of mutant alpha1 antitrypsin Z.
PMID 10636901
It is now well known that the addition and trimming of oligosaccharide side chains during post-translational modification play an important role in determining the fate of secretory, membrane, and lysosomal glycoproteins. Recent studies have suggested that trimming of oligosaccharide side chains also plays a role in the degradation of misfolded glycoproteins as a part of the quality control mechanism of the endoplasmic reticulum (ER). In this study, we examined the effect of several inhibitors of carbohydrate processing on the fate of the misfolded secretory protein alpha1 antitrypsin Z. Retention of this misfolded glycoprotein in the ER of liver cells in the classical form of alpha1 antitrypsin (alpha1-AT) deficiency is associated with severe liver injury and hepatocellular carcinoma and lack of its secretion is associated with destructive lung disease/emphysema. The results show marked alterations in the fate of alpha1 antitrypsin Z (alpha1-ATZ). Indeed, one glucosidase inhibitor, castanospermine (CST), and two mannosidase inhibitors, kifunensine (KIF) and deoxymannojirimycin (DMJ), mediate marked increases in secretion of alpha1-ATZ by distinct mechanisms. The effects of these inhibitors on secretion have interesting implications for our understanding of the quality control apparatus of the ER. These inhibitors may also constitute models for development of additional drugs for chemoprophylaxis of liver injury and emphysema in patients with alpha1-AT deficiency.
Related Materials
Product #
Molecular Formula
Add to Cart
N-Methyl-1-deoxynojirimycin, ≥98%