EMAIL THIS PAGE TO A FRIEND
Leukemia
Leukemia cell lines resistant to cytosine arabinoside, cyclophosphamide or daunomycin are equally sensitive to the drug acetyldinaline as the parental cell line.
PMID 10637498
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0621
CI−994, ≥98% (HPLC), powder
C15H15N3O2