EMAIL THIS PAGE TO A FRIEND
Genomics
Molecular cloning and mapping of a novel ADAM gene (ADAM29) to human chromosome 4.
PMID 10644455
Abstract
Members of the ADAM family (type I integral membrane protein with a disintegrin and metalloprotease domain) have been implicated in many important biological processes involving cell-cell and cell-matrix interactions, such as fertilization and myoblast fusion. We report here the cDNA sequence of a novel human ADAM gene (ADAM29) that contains a putative fusion peptide. Northern blot analysis revealed that the mRNA of ADAM29 is highly expressed in the testis. By radiation hybrid panel mapping, the ADAM29 gene was assigned to human chromosome 4q34.2-qter.