EMAIL THIS PAGE TO A FRIEND
Journal of the Royal Society of Medicine
The puzzle of pink disease.
PMID 10645305
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
230405
Mercury(I) chloride, ACS reagent, ≥99.5%
Cl2Hg2