EMAIL THIS PAGE TO A FRIEND
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology
Cromolyn sodium or nedocromil in childhood asthma: does it matter?
PMID 10651767