EMAIL THIS PAGE TO A FRIEND
Human mutation
18 bp insertion/duplication with internal missense mutation in human hepatic lipase gene exon 3. Mutations in brief no. 181. Online.
PMID 10660332
Abstract
Human hepatic lipase (hHL) plays an important role in hydrolysis of triglycerides from plasma lipoproteins. The enzyme also hydrolyzes HDL2 lipids resulting in smaller HDL particles with a lower cholesterol content and properties similar to HDL3. hHL is localized in liver sinusoids, ovary and adrenal gland. These findings propose an influence on processing of cholesterol. Here we report an insertion mutation in exon 3 of hHL. The 18 bp duplication contains an additional internal point mutation (GenBank-Accession #AF037404). The female mutation carrier suffered from severe adiposity with total cholesterol of 291,6mg/dl, HDL-cholesterol of 55,3mg/dl, LDL-cholesterol of 206,8mg/dl and triglycerides of 80,8mg/dl. Following cloning of a PCR-amplified fragment the mutation was confirmed by cycle sequencing. Sequence analysis revealed an inserted repeat of 18 nucleotides. Furthermore the patient carries an additional missense mutation A-->G at nucleotide 9 of the repeat which results in an amino acid exchange from Ile-->Val at codon 4 of the repeat. These data enable us to report the insertion of HisTyrThrValArgVal which might be responsible for the moderate shift in lipid metabolism of the heterozygous patient.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
62287 Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
L4777
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
62288 Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
62301 Lipase from Aspergillus niger, powder (fine), ~200 U/g
L4277 Lipase from Aspergillus oryzae, ≥20,000 U/g
62285 Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
75577 Lipase from Burkholderia sp., lyophilized powder, slightly beige, ~12 U/mg
02569 Lipase from Candida antarctica, lyophilized, powder, beige, ~0.3 U/mg
62302 Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
L1754 Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
62316 Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
L8525 Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
L3170
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
L8906 Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
L9031 Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
62298 Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
L0382 Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
62309 Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
28602 Lipase from Pseudomonas fluorescens, powder, slightly beige, ≥160 U/mg
L9518
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
62310 Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
L3001 Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg protein
62306 Lipase from wheat germ, ~0.1 U/mg
73940 Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg