EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
A novel, potent, and selective 5-HT(7) antagonist: (R)-3-(2-(2-(4-methylpiperidin-1-yl)ethyl)pyrrolidine-1-sulfonyl) phen ol (SB-269970).
PMID 10669560
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7389
SB-269970 hydrochloride, powder, ≥98% (HPLC)
C18H28N2O3S · HCl