EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Novel 2,2-dioxide-4,4-disubstituted-1,3-H-2,1,3-benzothiadiazines as non-nucleoside reverse transcriptase inhibitors.
PMID 10673109
Abstract
Benzothiadiazine non-nucleoside reverse transcriptase inhibitors (NNRTIs) of HIV have been synthesized via a novel process to afford active inhibitors, with the most potent compound exhibiting an IC90 = 180 nM in a whole cell assay. The 2,2-dioxide-1H-2,1,3-benzothiadiazine ring system was constructed in one step from 2-amino-5-chlorobenzonitrile.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
174335
2-Amino-5-chlorobenzonitrile, 98%
C7H5ClN2