EMAIL THIS PAGE TO A FRIEND
American journal of respiratory and critical care medicine
Leukotrienes in cardiovascular diseases.
PMID 10673238
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0411
BAY-X-1005, ≥98% (HPLC)
C23H23NO3