EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Enantiomer separation of alpha-ionone using gas chromatography with cyclodextrin derivatives as chiral stationary phases.
PMID 10674942
Abstract
The gas chromatographic enantiomer separation of alpha-ionone was studied with three different chiral stationary phases using as chiral selectors: (1) heptakis(2,3,6-tri-O-methyl)-beta-cyclodextrin, dissolved in polysiloxane PS-086, (2) octakis(2,6-di-O-pentyl-3-O-trifluoroacetyl)-gamma-cyclodextrin and (3) octakis(2,6-di-O-pentyl-3-O-butanoyl)-gamma-cyclodextrin, both dissolved in polysiloxane SE-54. The influence of the concentration of the chiral selector in the polysiloxane, coated on Chromosorb P AW-DMCS 80-100 mesh, is described and discussed, as well as the effect of Chromosorb loading. The feasibility of the preparative gas chromatographic separation of the enantiomers of alpha-ionone is considered; in order to provide a term of comparison, the estimated performances are compared with those achieved in the separation of the enantiomers of the inhalation anaesthetic enflurane.
Related Materials