EMAIL THIS PAGE TO A FRIEND
The Journal of veterinary medical science
Apoptosis and cell proliferation in rat hepatocytes induced by barbiturates.
PMID 10676885
Abstract
To examine the effect on cell population in hepatocytes of phenobarbital (PB) and other barbiturates, PB, allobarbital (ALB), barbital sodium (BS) and barbituric acid (BA) were given orally to male rats for 7 consecutive days. Although there was no apparent change in non-promoting BA, hepatomegaly was induced by PB, BS and ALB, which are promoters of hepatocarcinogenesis. In PB- and BS-treated livers, hepatomegaly was attributable to hepatocyte proliferation and enzyme induction. In ALB-treated liver, it was attributable to enzyme induction. The level of cell proliferation was reduced to less than the control values following withdrawal of PB, ALB and BS. It seemed that the degree of suppression of cell proliferation following withdrawal of these compounds correlated to the degree of cell proliferation (PB>BS>ALB) during treatment. In PB-treated liver, apoptosis was induced during treatment, serving to eliminate the excess of hepatocytes. This suggests that short-term administration of PB neither induced suppression of apoptosis nor disturbed homeostasis of hepatocyte populations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5761
5,5-Diallylbarbituric acid, analytical standard
C10H12N2O3