EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Biomedical sciences and applications
3-Hydroxyanthranilic acid-derived compounds formed through electrochemical oxidation.
PMID 10677004
Abstract
3-Hydroxyanthranilic acid (3-HAA)-derived oxidation products were analyzed using high-performance liquid chromatography with an electrochemical reactor and diode array detection and high-performance liquid chromatography with an electrochemical reactor and UV detection coupled with mass spectrometry. In addition to 3-HAA dimers such as cinnabarinic acid (CA), 6-amino-3-[(2-carboxy-6-hydroxyphenyl)amino]-2,5-dioxo-1,3-cyclohexadien e-1-carboxylic acid and 4,7-diamino-8-hydroxy-6H-dibenzo[a,d]pyran-6-one-3-carboxylic acid, a 3-HAA trimer and a 3-HAA tetramer were also detected and identified based on their electrospray ionization mass spectra and their UV-visible spectra. These five oxidation products were also detected on the elution profiles of high-performance liquid chromatography-diode array detection analyses for the reaction mixtures of the auto-oxidation of 3-HAA, of 3-HAA with potassium ferricyanide, of 3-HAA with horseradish peroxidase and hydrogen peroxide, and of 3-HAA with superoxide dismutase (SOD). 4,7-Diamino-8-hydroxy-6H-dibenzo[a,d]pyran-6-one-3-carboxylic acid was predominant in the auto-oxidation, in the reaction of 3-HAA with horseradish peroxidase and hydrogen peroxide, and in the electrochemical oxidation of 3-HAA at an applied potential of 0.0 V. On the other hand, CA, the 3-HAA trimer and the 3-HAA tetramer were predominant in the reaction of 3-HAA with K3[Fe(CN)6] and in the electrochemical oxidation of 3-HAA at an applied potential of 1.0 V.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0096
Cinnabarinic Acid, ≥98% (HPLC)
C14H8N2O6