EMAIL THIS PAGE TO A FRIEND
International journal of molecular medicine
Cancer cell selectivity of 5-iodo-6-aminobenzopyrone (INH2BP) and methyl-3,5-diiodo-4(4'-methoxyphenoxy) benzoate (DIME).
PMID 10677569
Abstract
The cellular pharmacologic actions, as measured by cell killing, of INH2BP, DIME and INO2BA (+ BSO) were determined in three types of cancer cells and compared to their action on quiescent confluent human foreskin fibroblast (HSF) and pre-confluent growing fibroblasts. The confluent HSF cells were completely refractory to the action of INH2BP and DIME, but were killed by INO2BA (+ BSO). Proliferating HSF and all three tumor cell types were killed by all three drugs. The apparent in vivo tumor specificity of INH2BP and DIME is explained by preferential cell cycle dependent selective drug uptake into tumor cells and by drug metabolism that reverses drug action in less vigorously cycling normal cells. The covalent binding of iodonitrosobenzamide (formed from INO2BA) and its toxicity are regulated by the concentration of GSH, and exhibit no cell cycle selectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2784
INH2BP, ≥98% (HPLC), solid
C9H6INO2