EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Cloning and characterization of a novel adaptor protein, CIN85, that interacts with c-Cbl.
PMID 10679202
Abstract
The c-Cbl protooncogene product is a prominent substrate of protein tyrosine kinases and is rapidly tyrosine-phosphorylated upon stimulation of a wide variety of cell-surface receptors. We have identified a novel c-Cbl-interacting protein termed CIN85 with a molecular mass of 85 kDa which shows similarity to adaptor proteins, CMS and CD2AP. CIN85 mRNA is expressed ubiquitously in normal human tissues and cancer cell lines analyzed. CIN85 was basally associated with c-Cbl. For interaction of CIN85 with c-Cbl, the second SH3 domain of CIN85 was shown to serve as a central player. The CIN85-c-Cbl association was enhanced shortly after stimulation of 293 cells with epidermal growth factor (EGF) and gradually diminished to a basal level, which correlated with a tyrosine phosphorylation level of c-Cbl. Our results suggest that CIN85 may play a specific role in the EGF receptor-mediated signaling cascade via its interaction with c-Cbl.