EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Selective involvement of cytochrome P450 2D subfamily in in vivo 4-hydroxylation of amphetamine in rat.
PMID 10681381
Abstract
The cytochrome P450 (P450) 2D subfamily catalyzes ring hydroxylation of amphetamines. We tested the hypothesis that P450 2D is selectively involved in amphetamine 4-hydroxylation. Urinary elimination of 4-hydroxyamphetamine and amphetamine was determined in male Sprague-Dawley rats pretreated with P450 inducers and inhibitors. The urinary 24-h metabolic ratio (amphetamine/4-hydroxyamphetamine) was not affected by the inducers 3-methylcholanthrene, isosafrole, phenobarbital, ethanol, pregnenolone-alpha-carbonitrile, and clofibrate. Isosafrole did, however, increase amphetamine elimination along with urine volume. Urinary elimination of 4-hydroxyamphetamine was significantly decreased by, and the metabolic ratio increased by, the inhibitors 1-aminobenzotriazole, CCl(4), quinidine, quinine, and primaquine. Diallyl sulfide and troleandomycin had no effect. In rat liver microsomes primaquine was shown to be an inhibitor of 2D activity. Urine 4-hydroxyamphetamine content correlated strongly (r(2) = 0. 989) with microsomal P450 2D activity in parallel-treated rats. These studies also substantiate that 4-hydroxylation of amphetamine is selectively performed by the P450 2D subfamily in the rat.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
329606
Isosafrol, mixture of cis and trans
C10H10O2