EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Arabidopsis cytochrome P450s that catalyze the first step of tryptophan-dependent indole-3-acetic acid biosynthesis.
PMID 10681464
Abstract
Plants synthesize numerous secondary metabolites that are used as developmental signals or as defense against pathogens. Tryptophan (Trp)-derived secondary metabolites include camalexin, indole glucosinolates, and indole-3-acetic acid (IAA); however, the steps in their synthesis from Trp or its precursors remain unclear. We have identified two Arabidopsis cytochrome P450s (CYP79B2 and CYP79B3) that can convert Trp to indole-3-acetaldoxime (IAOx), a precursor to IAA and indole glucosinolates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1002
Acetaldehyde oxime, mixture of syn and anti, 99%
C2H5NO