EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Identification and characterization of a novel hepatic canalicular ATP diphosphohydrolase.
PMID 10681547
Abstract
We have identified and characterized a novel ATP diphosphohydrolase (ATPDase) with features of E-type ATPases from porcine liver. Immunoblotting with a specific monoclonal antibody to this ectoenzyme revealed high expression in liver with lesser amounts in kidney and duodenum. This ATPDase was localized by immunohistochemistry to the bile canalicular domain of hepatocytes and to the luminal side of the renal ductular epithelium. In contrast, ATPDase/cd39 was detected in vascular endothelium and smooth muscle in these organs. We purified the putative ATPDase from liver by immunoaffinity techniques and obtained a heavily glycosylated protein with a molecular mass estimated at 75 kDa. This enzyme hydrolyzed all tri- and diphosphonucleosides but not AMP or diadenosine polyphosphates. There was an absolute requirement for divalent cations (Ca(2+) > Mg(2+)). Biochemical activity was unaffected by sodium azide or other inhibitors of ATPases. Kinetic parameters derived from purified preparations of hepatic ATPDase indicated V(max) of 8.5 units/mg of protein with apparent K(m) of 100 microM for both ATP or ADP as substrates. NH(2)-terminal amino acid sequencing revealed near 50% identity with rat liver lysosomal (Ca(2+)-Mg(2+))-ATPase. The different biochemical properties and localization of the hepatic ATPDase suggest pathophysiological functions that are distinct from the vascular ATPDase/cd39.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9128
5′-(4-Fluorosulfonylbenzoyl)adenosine hydrochloride
C17H16FN5O7S · HCl