EMAIL THIS PAGE TO A FRIEND
Molecular pathology : MP
A colorectal cell line with alterations in E-cadherin and epithelial biology may be an in vitro model of colitis.
PMID 10694944
Abstract
It has been shown previously in ulcerative colitis tissue that E-cadherin can occasionally be mutated in the extracellular domain early in neoplastic progression. E-cadherin is known to maintain differentiation and inhibits invasion in vivo. To assess the mechanisms by which such dysfunction occurs. Four human colorectal cancer cell lines, HCA-7 colonies 1, 3, 6, and 30, derived from a single heterogeneous colorectal cancer were studied. The HCA-7 cell line has p53 mutations and a random errors of replication "positive" phenotype, as is seen in early colitis associated cancers or hereditary nonpolyposis coli cancer (HNPCC). Cell lines 6 and 30 expressed E-cadherin abundantly and this correlated positively with their degree of differentiation and organisation; however, both cell lines had loss of heterozygosity of E-cadherin. Interestingly, E-cadherin production was downregulated in the poorly differentiated cell line 1, and this was associated with major chromosomal rearrangements of 16q. This cell line also had a mutation in the homophilic binding domain of exon 4, which was associated with disaggregation by low titres of a function blocking antibody, and an invasive phenotype. These multiple biological alterations further characterise the complex association that E-cadherin has with tumour heterogeneity and suggest that this series of cell lines may be a useful model of colitis associated or HNPCC associated tumorigenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
HCA-7, 06061902