EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Involvement of acyl coenzyme A oxidase isozymes in biotransformation of methyl ricinoleate into gamma-decalactone by Yarrowia lipolytica.
PMID 10698800
Abstract
We reported previously on the function of acyl coenzyme A (acyl-CoA) oxidase isozymes in the yeast Yarrowia lipolytica by investigating strains disrupted in one or several acyl-CoA oxidase-encoding genes (POX1 through POX5) (H. Wang et al., J. Bacteriol. 181:5140-5148, 1999). Here, these mutants were studied for lactone production. Monodisrupted strains produced similar levels of lactone as the wild-type strain (50 mg/liter) except for Deltapox3, which produced 220 mg of gamma-decalactone per liter after 24 h. The Deltapox2 Deltapox3 double-disrupted strain, although slightly affected in growth, produced about 150 mg of lactone per liter, indicating that Aox2p was not essential for the biotransformation. The Deltapox2 Deltapox3 Deltapox5 triple-disrupted strain produced and consumed lactone very slowly. On the contrary, the Deltapox2 Deltapox3 Deltapox4 Deltapox5 multidisrupted strain did not grow or biotransform methyl ricinoleate into gamma-decalactone, demonstrating that Aox4p is essential for the biotransformation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R8750
Methyl ricinoleate, ≥99% (GC)
C19H36O3
83916
Methyl ricinoleate, analytical standard
C19H36O3