EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Densitometric determination of impurities in drugs: Part IV. Determination of N-(4-aminobenzoyl)-L-glutamic acid in preparations of folic acid.
PMID 10704002