EMAIL THIS PAGE TO A FRIEND
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology
Nedocromil sodium: a review of the evidence for a dual mechanism of action.
PMID 10718842