EMAIL THIS PAGE TO A FRIEND
Arzneimittel-Forschung
Antioxidant properties of essential oils. Possible explanations for their anti-inflammatory effects.
PMID 10719616
Abstract
Pathogenesis and symptoms of inflammatory processes are accompanied and/or initiated by the production of reactive oxygen species (ROS). The effects of essential oils on these processes have been studied with the aid of biochemical model reactions simulating these pathological events. It can be shown that Myrtol Standardized and Eucalyptus oil ameliorate inflammatory processes by interacting with aggressive oxygen radicals of the OH.-type and interfere with leukocyte activation. These activities partially allow attenuation of oxidative attack and damage introduced by infections or environmental impacts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K6000
α-Keto-γ-(methylthio)butyric acid sodium salt, ≥97%
C5H7NaO3S