EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Copper has differential effect on prion protein with polymorphism of position 129.
PMID 10720484
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB5058
Anti-Prion Protein Antibody, NT, a.a. 78-97, serum, clone, Chemicon®