EMAIL THIS PAGE TO A FRIEND
Toxicology letters
Glutathione S-transferase: genetics and role in toxicology.
PMID 10720752
Abstract
The glutathione S-transferases (GST) are a supergene family of dimeric, enzymes that catalyse the conjugation of glutathione (GSH) to a variety of electrophiles including arene oxides, unsaturated carbonyls, organic halides and other substrates. Their importance is suggested by the finding that GST enzymes are expressed in probably all life forms. In humans, polymorphism in GST genes has been associated with susceptibility to various diseases though some recent data indicate that these genotypes modify disease phenotype. Thus, GST genotypes alone and in combination have been linked with clinical outcome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GS39 GST T2-2, Recombinant Rat