EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Different catalytically competent arrangements of arachidonic acid within the cyclooxygenase active site of prostaglandin endoperoxide H synthase-1 lead to the formation of different oxygenated products.
PMID 10722687
Abstract
Arachidonic acid is converted to prostaglandin G(2) (PGG(2)) by the cyclooxygenase activities of prostaglandin endoperoxide H synthases (PGHSs) 1 and 2. The initial, rate-limiting step is abstraction of the 13-proS hydrogen from arachidonate which, for PGG(2) formation, is followed by insertion of O(2) at C-11, cyclization, and a second O( 2) insertion at C-15. As an accompaniment to ongoing structural studies designed to determine the orientation of arachidonate in the cyclooxygenase site, we analyzed the products formed from arachidonate by (a) solubilized, partially purified ovine (o) PGHS-1; (b) membrane-associated, recombinant oPGHS-1; and (c) a membrane-associated, recombinant active site mutant (V349L oPGHS-1) and determined kinetic values for formation of each product. Native forms of oPGHS-1 produced primarily PGG(2) but also several monohydroxy acids, which, in order of abundance, were 11R-hydroxy-5Z, 8Z,12E,14Z-eicosatetraenoic acid (11R-HETE), 15S-hydroxy-5Z,8Z,11Z, 13E-eicosatetraenoic acid (15S-HETE), and 15R-HETE. V349L oPGHS-1 formed primarily PGG(2), 15S-HETE, and 15R-HETE but only trace amounts of 11R-HETE. With native enzyme, the K(m) values for PGG(2), 11-HETE, and 15-HETE formation were each different (5.5, 12.1, and 19.4 microM, respectively); similarly, the K(m) values for PGG(2) and 15-HETE formation by V349L oPGHS-1 were different (11 and 5 microM, respectively). These results establish that arachidonate can assume at least three catalytically productive arrangements within the cyclooxygenase site of oPGHS-1 leading to PGG(2), 11R-HETE, and 15S-HETE and/or 15R-HETE, respectively. IC(50) values for inhibition of formation of the individual products by the competitive inhibitor, ibuprofen, were determined and found to be the same for a given enzyme form (i.e. 175 microM for oPGHS-1 and 15 microM for V349L oPGHS-1). These latter results are most simply rationalized by a kinetic model in which arachidonate forms various catalytically competent arrangements only after entering the cyclooxygenase active site.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7742
Prostaglandin G2, ≥95% (HPLC), acetone solution
C20H32O6