EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Electron paramagnetic resonance study of V2O5 deactivation in the course of catalytic oxidation of SO2 to SO3.
PMID 10727148
Abstract
The influence of reagents interactions with V2O5 catalyst on its activity in the reaction of SO2 oxidation has been investigated by electron paramagnetic resonance (EPR) method. It was found that the deactivation of V2O5 is caused by the diminishing of the number of adsorption centers able to interact with reagent molecules and with the creation of elements of the VOSO4 phase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
227692
Sulfur trioxide, contains stabilizer, ≥99%
O3S