EMAIL THIS PAGE TO A FRIEND
Bioorganicheskaia khimiia
[Synthesis of novel fluorescent 9,10-bisphenylethynylanthracene-derived pseudonucleoside and its introduction into oligonucleotides].
PMID 10734554
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45788
9,10-Diphenylanthracene, analytical standard
C26H18
D205001
9,10-Diphenylanthracene, 97%
C26H18