EMAIL THIS PAGE TO A FRIEND
Glycoconjugate journal
A new technique distinguishing alpha2-3 sialyl linkage from alpha2-6 linkage in sialyllactoses and sialyl-N-acetyllactosamines by post-source decay fragmentation method of MALDI-TOF mass spectrometry.
PMID 10737323
Abstract
Alpha2-3 and alpha2-6 sialyl linkage types of sialyllactoses and sialyl-N-acetyllactosamines were analyzed by post-source decay (PSD) fragmentation method using matrix-assisted laser desorption/ionization time-of-flight (MALDI-TOF) mass spectrometry. A new matrix of norharmane was suited for the MALDI-TOF measurements of sialyl oligosaccharides. The fragment ions B1 produced by the cleavage of alpha2-3 sialyl linkages indicate much higher intensity than those produced by the cleavage of alpha2-6 sialyl linkages in sialyllactoses and sialyl-N-acetyllactosamines. Thus, alpha2-3 sialyl linkages cleave much easier than alpha2-6 sialyl linkages in MALDI-PSD fragmentation method. These results suggest that the new techniques using PSD fragmentation of MALDI-TOF mass spectrometry enables us to distinguish alpha2-3 sialyl linkage from alpha2-6 linkage in sialyl oligosaccharides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8681
3′-Sialyllactose, from bovine milk or colostrum, ≥97% (HPLC)
C23H39NO19