EMAIL THIS PAGE TO A FRIEND
Research communications in molecular pathology and pharmacology
A prostaglandin E1 analog, OP-1206, alleviates 5-fluorouracil-induced injury of rat small intestine.
PMID 10741375
Abstract
5-Fluorouracil (5-FU) often causes the gastrointestinal toxicity, including enterocolitis. We investigated effects of OP-1206 (17S, 20-dimethyl-trans-delta2-prostaglandin E1) on 5-FU-induced leukocyte infiltration and epithelial barrier dysfunction of rat small intestine. Myeloperoxidase (MPO) activity of the small intestine was assayed as an index of leukocyte infiltration. Intestinal epithelial permeability was determined by the small intestinal absorption of a paracellular permeation marker, fluorescein isothiocyanate-labeled dextran (molecular weight; 4,400) (FD-4) using the in situ closed intestinal loop technique. The MPO activity and FD-4 permeation were significantly increased by the administration of 5-FU to rats for 4 days, while on the coadministration of 5-FU and OP-1206, they were similar to those of control rats treated with saline solution alone, respectively. These observations indicate that OP-1206 reduced the leukocyte infiltration and the change in epithelial permeability of rat small intestine induced by 5-FU.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6538
Limaprost, ≥99%, crystalline
C22H36O5