EMAIL THIS PAGE TO A FRIEND
Seminars in ophthalmology
Intraocular gas tamponades.
PMID 10749310
Abstract
Intraocular gas tamponades are an important part of vitreoretinal surgery and have become a standard of care. The use of intraocular gas began in 1911. Modem use of intraocular gases as a surgical adjunct is continuously broadening. Today, sulfur hexafluoride and perfluoropropane are increasingly being used as intraocular gas tamponades for a wide range of complicated vitreoretinal diseases. This article reviews the types and function of commonly used gases, their indications and efficacy in certain types of vitreoretinal diseases, and their complications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295736
Carbon tetrafluoride, 99.9%
CF4