EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Enzymatic desymmetrization of a meso polyol corresponding to the C(19)-C(27) segment of rifamycin S.
PMID 10750492
Abstract
The stereoselective acylation of meso polyol 2 by vinyl acetate (solvent and acyl donor) in the presence of porcine pancreas lipase gave the corresponding monoester 5 in good yield (76%) and high enantiomeric purity (ee > 98%). The enzymatic reaction was also highly regioselective for a primary alcohol end group, and the two unprotected secondary alcohols were not involved. Compound 5 corresponds to the C(19)-C(27) fragment of rifamycin S.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0950000 Rifamycin S, European Pharmacopoeia (EP) Reference Standard
C37H45NO12