EMAIL THIS PAGE TO A FRIEND
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
The discrepancy between the binding affinity of PTH (1-34) and RS 66271 is explained by interaction of the PTH/PTHrP receptor with G-protein.
PMID 10750577